Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Dividers for Slide Boxes

Dividers for Slide Boxes

Divider for slides and coverslips of special sizes, bianco, PS, 16,5×76×5 mm, 15
Codice catalogo GLWAZK15
Nome del fornitore:   GLW
Dividers for Slide Boxes
Codice catalogo GLWAZK15
Nome del fornitore:  GLW
Dividers for Slide Boxes
Dividers for Slide Boxes

Divider for slides and coverslips of special sizes, bianco, PS, 16,5×76×5 mm, 15

Price
SFr. 6.50
Log in per disponibilità
1 * 1 Un.
Product Details & Documents

These slide box dividers are designed to store different slides and coverslips of various dimensions.

  • Adjustable length, pluggable into slide boxes
  • Can be used in in freezer down to −90 °C

For use with standard and special sized slides.

Attenzione: Not for use in autoclaves.

Specifiche
  • N. di vetrini:
    15
  • Colore:
    bianco
  • Descrizione:
    Divider for slides and coverslips of special sizes
  • Materiale:
    PS
  • Seq:
    01
  • L×P×A:
    16,5×76×5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.