Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cryobank™ Preservation System

Cryobank™ Preservation System

Storage box for 81 vials each, 1
Codice catalogo MAST291690
Nome del fornitore:   MAST DIAGNOSTICA
Cryobank™ Preservation System
Codice catalogo MAST291690
Nome del fornitore:  MAST DIAGNOSTICA
Numero CAS:  
Cryobank™ Preservation System
Cryobank™ Preservation System

Storage box for 81 vials each, 1

Numero CAS:  
Price
SFr. 32.80
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Cryobank™ is a preservation system with a high recultivation rate. Colour coded tubes make it easy to find the mircroorganisms.

  • Quick
  • Efficient
  • Reliable
Specifiche

Specifiche

  • Conf:
    1
  • Descrizione:
    Storage box for 81 vials each

Specifiche