Orders LinkContactUsLinkComponent
Computers for Laboratory Equipment
Computers for Laboratory Equipment
PC Dell Optiplex XE4 Tower - Processor Intel Core i7-12700 (8+4C - Up to 4,90 GHz); 16 GB DDR4 Memory - 1TB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 11 IoT Ent. 2024 LTSC Value license operating system
Codice catalogo 906-0250
Nome del fornitore:   COLLECTION PREFIX HPLC
Computers for Laboratory Equipment
Codice catalogo 906-0250
Nome del fornitore:  COLLECTION PREFIX HPLC
Computers for Laboratory Equipment
Computers for Laboratory Equipment
PC Dell Optiplex XE4 Tower - Processor Intel Core i7-12700 (8+4C - Up to 4,90 GHz); 16 GB DDR4 Memory - 1TB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 11 IoT Ent. 2024 LTSC Value license operating system
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
  • Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
  • Compact footprint with integrated PC-monitor solutions
  • Pre-installed language packs
  • Keyboards with all common European language layouts
  • Compatible with main CDS (Chromatography Data Systems)
Specifiche
  • Descrizione:
    PC Dell Optiplex XE4 Tower - Processor Intel Core i7-12700 (8+4C - Up to 4,90 GHz); 16 GB DDR4 Memory - 1TB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 11 IoT Ent. 2024 LTSC Value license operating system
  • Seq:
    09
Complementary Products
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • 906-0219
    Dell™ mouse, bundled with Dell™ keyboard, DE
    Log in per disponibilità
    1 * 1 ST - 32.40 SFr.
  • 906-0211
    Dell™ mouse, bundled with Dell™ keyboard, FR
    Log in per disponibilità
    1 * 1 ST - 32.40 SFr.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.