Orders LinkContactUsLinkComponent
Cell Counting Kit 8 (CCK-8)
Cell Counting Kit 8 (CCK-8)
10000 tests, 10.000 Tests
Codice catalogo AATB22773
Nome del fornitore:   AAT Bioquest
Cell Counting Kit 8 (CCK-8)
Codice catalogo AATB22773
Nome del fornitore:  AAT Bioquest
Numero CAS:  
Cell Counting Kit 8 (CCK-8)
10000 tests, 10.000 Tests
Numero CAS:  
Price
SFr. 1’890.00
Log in per disponibilità
1 * 10.000 Tests
Product Details & Documents
Designed for consistent and dependable results. Cell Counting Kit 8 (CCK-8) is a colorimetric assay used to quantitate cell proliferation, cell viability and cytotoxicity
  • Improved water solubility compared to MTT assays
  • Excellent stability allowing for long assays (24 to 48 hours)
  • Single-component, mix-and-read format
CCK-8 is a colorimetric assay that relies on production of formazan from WST-8, catalysed by endogenous cellular dehydrogenase in the presence of NADPH from metabolically active cells.
 : Shipped with blue ice.
 : Research use only.
Specifiche
Specifiche
  • Conf:
    10.000 Tests
  • Test per kit:
    10000 tests