Cell Counting Kit 8 (CCK-8)
10000 tests, 10.000 Tests
Codice catalogo AATB22773
Nome del fornitore: AAT Bioquest
Vai a
Product Details & Documents
Designed for consistent and dependable results. Cell Counting Kit 8 (CCK-8) is a colorimetric assay used to quantitate cell proliferation, cell viability and cytotoxicity
- Improved water solubility compared to MTT assays
- Excellent stability allowing for long assays (24 to 48 hours)
- Single-component, mix-and-read format
CCK-8 is a colorimetric assay that relies on production of formazan from WST-8, catalysed by endogenous cellular dehydrogenase in the presence of NADPH from metabolically active cells.
: Shipped with blue ice.
: Research use only.
Specifiche
Specifiche
- Conf:10.000 Tests
- Test per kit:10000 tests