Orders LinkContactUsLinkComponent
Cell Counting Kit 8 (CCK-8)
Cell Counting Kit 8 (CCK-8)
1000 tests, 1.000 Tests
Codice catalogo AATB22772
Nome del fornitore:   AAT Bioquest
Cell Counting Kit 8 (CCK-8)
Codice catalogo AATB22772
Nome del fornitore:  AAT Bioquest
Numero CAS:  
Cell Counting Kit 8 (CCK-8)
1000 tests, 1.000 Tests
Numero CAS:  
Price
SFr. 315.00
Log in per disponibilità
1 * 1.000 Tests
Product Details & Documents
Designed for consistent and dependable results. Cell Counting Kit 8 (CCK-8) is a colorimetric assay used to quantitate cell proliferation, cell viability and cytotoxicity
  • Improved water solubility compared to MTT assays
  • Excellent stability allowing for long assays (24 to 48 hours)
  • Single-component, mix-and-read format
CCK-8 is a colorimetric assay that relies on production of formazan from WST-8, catalysed by endogenous cellular dehydrogenase in the presence of NADPH from metabolically active cells.
Informazioni per l'ordine: Shipped with blue ice.
Attenzione: Research use only.
Specifiche
Specifiche
  • Conf:
    1.000 Tests
  • Test per kit:
    1000 tests
Specifiche
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.