[EN]5 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM040268L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
[EN]5 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM040268L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Kit analitici, COD in cuvetta per Spectroquant®, Supelco®
Supplier: Merck
Kit analitici, test cianuri per Spectroquant®, Supelco®
Supplier: Merck
Kit analitici, tensioattivi, per Spectroquant®, Supelco®
Supplier: Merck
Kit colorimetrici, cianuri, MColortest™, Supelco®
Supplier: Merck
Kit analitici, nitrati per Spectroquant®, Supelco®
Supplier: Merck
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
OXALIC ACID 0.005M (0.01N) 17034 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Sodio idrossido, anidro ≥98%, in pastiglie, per analisi
Supplier: Honeywell Chemicals
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY