Accessories
Temperature probe made of stainless steel, PTFE-coated, with sensor PT100
Catalog # KRUEPRT-T
Supplier: A.KRÜSS Optronic
Jump to:
Product Details & Documents
Specifications
- Descrizione:Temperature probe made of stainless steel, PTFE-coated, with sensor PT100
- Seq:04