Mixed Anions Standard - 7 components: 1000mg/l each of Fluorides (F-) Chlorides (Cl-) Nitrites (NO2-) Bromides (Br-) Nitrates (NO3-) Sulphates (SO42-) Phosphates (PO43-) in Water 17034 and ISO 17025 Conservation 36 months. Store under nor 1 * 500 mL
Codice catalogo CMPL1521.1K.W.L5
Nome del fornitore: CUSTOM MADE CHEMICALS LAB