Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Enriched Foam™ Hand Washes

Enriched Foam™ hand wash for Manual Foam 400 ml dispenser, 400 ml
  RUBBRVU9001
Enriched Foam™ Hand Washes
  RUBBRVU9001
Enriched Foam™ Hand Washes
Enriched Foam™ Hand Washes

Enriched Foam™ hand wash for Manual Foam 400 ml dispenser, 400 ml

 
7.60 SFr.
   
1 * 1 ST
 

Light, floral, and aromatic fragranced formula with enriched moisturisers to help prevent skin dryness.

  • Hygienically sealed refill eliminates the potential for contamination
  • Dermatologically tested and enriched with skin conditioners
  • High capacity refills deliver up to 3250 hand washes for best cost in use
  • Appeals to all, ideal for high traffic environments
 
  • Capacità:
    400 ml
  • Descrizione:
    Enriched Foam™ hand wash for Manual Foam 400 ml dispenser
  • Seq:
    04
Complementary Products

 

  •  
    Colore
    Descrizione
    Capacità
     
     
  • RUBBFG450017
    Bianco/grigio
    Manual foam dispenser
    800 ml
       
    1 * 1 ST - SFr. 59.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.