Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Enriched Foam™ Hand Washes

Enriched Foam™ hand wash for FLEX™ 1300 ml dispenser, 1300 ml
  RUBB3486615
Enriched Foam™ Hand Washes
  RUBB3486615
Enriched Foam™ Hand Washes
Enriched Foam™ Hand Washes

Enriched Foam™ hand wash for FLEX™ 1300 ml dispenser, 1300 ml

 
SFr. 21.70
   
1 * 1 ST
 

Light, floral, and aromatic fragranced formula with enriched moisturisers to help prevent skin dryness.

  • Hygienically sealed refill eliminates the potential for contamination
  • Dermatologically tested and enriched with skin conditioners
  • High capacity refills deliver up to 3250 hand washes for best cost in use
  • Appeals to all, ideal for high traffic environments
 
  • Capacità:
    1300 ml
  • Descrizione:
    Enriched Foam™ hand wash for FLEX™ 1300 ml dispenser
  • Seq:
    05
Complementary Products

 

  •  
    Colore
    Descrizione
    Capacità
     
     
  • RUBBFG450017
    Bianco/grigio
    Manual foam dispenser
    800 ml
       
    1 * 1 ST - SFr. 59.00