Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]2,4-TRIMETHYLBENZENE 1 * 5 mL
Catalog # CMPLSB15.5000
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
[EN]2,4-TRIMETHYLBENZENE 1 * 5 mL
Catalog # CMPLSB15.5000
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Standard di conducibilità, Certipur®, Supelco®
Supplier: Merck
Soluzioni tampone colorate Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM, Soluzioni tampone, 20 °C
Supplier: VWR Chemicals
Soluzioni tampone Certipur®, Supelco®
Supplier: Merck
Soluzione tampone, pronta all'uso
Supplier: WTW
Soluzioni standard di conducibilità
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Potassio cloruro 3 mol/l (3 N) in soluzione acquosa, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck