Orders LinkContactUsLinkComponent
Accessories for Stereomicroscope
Accessories for Stereomicroscope
Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
Codice catalogo ZEIS4354339401000
Nome del fornitore:   ZEISS
Accessories for Stereomicroscope
Codice catalogo ZEIS4354339401000
Nome del fornitore:  ZEISS
Accessories for Stereomicroscope
Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Descrizione:
    Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
  • Seq:
    01