Accessories for Stereomicroscope
Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
Codice catalogo ZEIS4354339401000
Nome del fornitore: ZEISS
Vai a
Product Details & Documents
Specifiche
- Descrizione:Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
- Seq:01