Plunger cans
Plunger Can, 2 l, 145×155×265 mm
Codice catalogo MSGA40098
Nome del fornitore: asecos
Vai a
Product Details & Documents
These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.
- Unused liquid flows back into the container
- Spring mounted plate includes flame barrier
- Shock resistant and explosion proof (PE models only)
- Cans with protector ring (stainless steel models only)
Specifiche
- Capacità:2 l
- Descrizione:Plunger Can
- L×P×A:145×155×265 mm



