ContactUsLinkComponent
Vinylchloride [CAS:75-01-4] 1974mg/l 1,1-Dichloroethene [CAS:75-35-4] 2003mg/l Chloroform [CAS:67-66-3] 2001mg/l 1,2-Dichloropropane [CAS: 78-87-5] 2001mg/l Toluene [CAS:108-88-3] 2004mg/l Ethylbenzene [CAS:1 4] 2025mg/l in Methanol 1 * 5 Ampoll
Codice catalogo CMPLRD1091501.5
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
Vinylchloride [CAS:75-01-4] 1974mg/l 1,1-Dichloroethene [CAS:75-35-4] 2003mg/l Chloroform [CAS:67-66-3] 2001mg/l 1,2-Dichloropropane [CAS: 78-87-5] 2001mg/l Toluene [CAS:108-88-3] 2004mg/l Ethylbenzene [CAS:1 4] 2025mg/l in Methanol 1 * 5 Ampoll
Codice catalogo CMPLRD1091501.5
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
Frequently Bought Together