Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
EasyFrit ES1210 EasySampler Filter Kit

EasyFrit ES1210 EasySampler Filter Kit

Filter kit for ES1210, FEP, 14,2 mm, 47 mm
Codice catalogo METB30811900
Nome del fornitore:   Mettler - Toledo
EasyFrit ES1210 EasySampler Filter Kit
Codice catalogo METB30811900
Nome del fornitore:  Mettler - Toledo
EasyFrit ES1210 EasySampler Filter Kit

Filter kit for ES1210, FEP, 14,2 mm, 47 mm

Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents

EasyFrit turns EasySampler into a selective solution phase sampling tool for data-rich collection throughout a controlled crystallisation.

  • HPLC ready samples
  • Eliminate user safety concerns
  • Avoid surface fouling
  • Transform and increase productivity
  • Never miss a reaction event
  • Representative and reproducible
  • Material: FEP, alloy C-276, C22 alloy

A unique self-flushing mechanism prevents surface fouling and allows unattended sampling over an elongated period under reaction conditions at scale.

Unattended, representative sampling: EasyFrit is a dynamically flushed in situ filter. Its unique mechanism minimizes the risk of surface fouling. It is compatible with all EasySampler 1210 probes.

Automated and unattended sampling: EasyFrit allows for 24/7 data-rich collection of solution-phase information throughout a controlled crystallization, preventing data blind spots.

Representative and reproducible: EasySampler samples from the same position into a fixed volume pocket for accurate analytical data.

Reactions difficult to sample: Easily manage reactions with high solid loading using automated filtration, sample capture, and dilution.

Informazioni per l'ordine: The kit contains one assembled filter and the installation tool.

Specifiche
  • Ø:
    14,2 mm
  • Descrizione:
    Filter kit for ES1210
  • Lunghezza:
    47 mm
  • Materiale:
    FEP
  • Seq:
    02
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Disponibilità
    Price
  • Tube set probe 210, PTFE tube, PEEK fittings; 4 pcs in a set
    Tube set probe 210, PTFE tube, PEEK fittings; 4 pcs in a set
    METB30247094
    Log in per disponibilità
    1 * 1 ST

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..