Orders LinkContactUsLinkComponent
EasyFrit ES1210 EasySampler Filter Kit
Filter for ES1210 assembled, — mm, —
  METB30811887
EasyFrit ES1210 EasySampler Filter Kit
  METB30811887
EasyFrit ES1210 EasySampler Filter Kit
Filter for ES1210 assembled, — mm, —
1 * 1 ST
EasyFrit turns EasySampler into a selective solution phase sampling tool for data-rich collection throughout a controlled crystallisation.
  • HPLC ready samples
  • Eliminate user safety concerns
  • Avoid surface fouling
  • Transform and increase productivity
  • Never miss a reaction event
  • Representative and reproducible
  • Material: FEP, alloy C-276, C22 alloy
A unique self-flushing mechanism prevents surface fouling and allows unattended sampling over an elongated period under reaction conditions at scale.
Unattended, representative sampling: EasyFrit is a dynamically flushed in situ filter. Its unique mechanism minimizes the risk of surface fouling. It is compatible with all EasySampler 1210 probes.
Automated and unattended sampling: EasyFrit allows for 24/7 data-rich collection of solution-phase information throughout a controlled crystallization, preventing data blind spots.
Representative and reproducible: EasySampler samples from the same position into a fixed volume pocket for accurate analytical data.
Reactions difficult to sample: Easily manage reactions with high solid loading using automated filtration, sample capture, and dilution.
 : The kit contains one assembled filter and the installation tool.
  • Ø:
    — mm
  • Descrizione:
    Filter for ES1210 assembled
  • Lunghezza:
  • Seq:
    01
Accessories
  •  
    Descrizione
  • METB30247094
    Tube set probe 210, PTFE tube, PEEK fittings; 4 pcs in a set
    1 * 1 ST