Human recombinant Lin28a (full-length)
200 µG
Catalog # ENZOENZPRT1720200
Supplier: Enzo Life Sciences
Human recombinant Lin28a (full-length)
Catalog # ENZOENZPRT1720200
Supplier: Enzo Life Sciences
CAS Number:
Jump to:
Product Details & Documents
Produced in E. coli.
Specifications
Specifications
- Conf:200 µG
- Peso molecolare:~24 kDa
- Cat. No.:ENZOENZPRT1720200
- Protein/peptide type:Recombinant
- Protein/peptide name:Lin28a (full-length)
- Purezza:>95% (SDS-PAGE)
- Protein synonyms:microRNA|cancer|Protein lin-28 homolog A|stem cells
- Purificazione:Purified by multi-step chromatography.
- Specie:Human
- Formulazione:Liquid. In 20 mM TRIS, pH 7,2, containing 200 mM sodium chloride and 5 mM diothiothreitol (DTT).
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



