Orders LinkContactUsLinkComponent
HPV type 6/11 probe, PATHO-GENE®
HPV type 6/11 probe, PATHO-GENE®
HPV type 6/11 probe, PATHO-GENE®, 1 mL
Codice catalogo ENZOENZ32885
Nome del fornitore:   Enzo Life Sciences
HPV type 6/11 probe, PATHO-GENE®
Codice catalogo ENZOENZ32885
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
HPV type 6/11 probe, PATHO-GENE®
HPV type 6/11 probe, PATHO-GENE®
HPV type 6/11 probe, PATHO-GENE®, 1 mL
Numero CAS:  
Price
SFr. 1’490.00
Log in per disponibilità
1 * 1 mL
Product Details & Documents
The PATHO-GENE® HPV type 6/11 probe is a mixture of biotin-labeled HPV 6 and HPV 11-specific probes in buffered formamide and hybridisation enhancers.
The ATHO-GENE® HPV type 6/11 probe employs a mixture of the HPV 6 and HPV 11-specific probes that detect HPV types that have been associated with benign lesions presenting as condylomas and mild dysplasia. The probe mixture is denatured and hybridised to the tissue section fixed and pretreated on the microscope slide. Specific hybridisation between the HPV DNA probe and DNA in the specimen is determined by the detection of biotin.
Specifiche
Specifiche
  • Conf:
    1 mL
  • Descrizione:
    HPV type 6/11 probe, PATHO-GENE®