Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
U-73122 ≥99% (by TLC)

U-73122 ≥99% (by TLC)

25 mg
Catalog # ENZOBMLST3910025
Supplier:   Enzo Life Sciences
U-73122 ≥99% (by TLC)
Catalog # ENZOBMLST3910025
Supplier:  Enzo Life Sciences
CAS Number:  112648-68-7
U-73122 ≥99% (by TLC)

25 mg

CAS Number:  112648-68-7
Price
SFr. 592.00
Log in for availability
1 * 25 mg
Product Details & Documents

Inhibitor of PLC activation

Inhibits agonist induced phospholipase C (PLC) activation (IC50=1-2.1 µM) in human platelets. It also inhibits agonist induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells. It is a useful tool to investigate receptor-mediated PI turnover in signal transduction. It is a potent inhibitor of human neutrophil adhesion to biological surfaces as well as adhesion-dependent granule exocytosis and oxidative burst. Inhibits 5-lipoxygenase in vivo and in vitro (IC50 = 30nM for recombinant enzyme and 2.4µM for PMNL homogenates).

Synonyms:  1-(6-(((8R,9S,13S,14S,17S)-3-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthren-17-yl)amino)hexyl)-1H-pyrrole-2,5-dione, 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione
  • CAS Number:112648-68-7
  • Formula:C₂₉H₄₀N₂O₃
  • MDL Number:MFCD00893825
  • Molecular Weight:464.65 g/mol
Specifications

Specifications

  • Conf:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.