Orders LinkContactUsLinkComponent
Phytosphingosine ≥98%
25 mg
  ENZOBMLSL1500025
Phytosphingosine ≥98%
  ENZOBMLSL1500025
 :  554-62-1
Phytosphingosine ≥98%
Phytosphingosine ≥98%
25 mg
 :  554-62-1
SFr. 482.00
1 * 25 mg
Ultra-pure
An endogenous sphingosine occurring in Saccharomyces cerevisiae. Inhibits FMLP-induced oxidant release by human neutrophils.
 :  (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
  •  :554-62-1
  •  :C₁₈H₃₉NO₃
  •  :MFCD00079232
  •  :317.51 g/mol
  • Conf:
    25 mg