AG-879 ≥99%
25 mg
ENZOBMLEI2580025
NGF receptor inhibitor
AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells.
: Tyrphostin AG 879, (E)-2-Cyano-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enethioamide
- :148741-30-4
- :C₁₈H₂₄N₂OS
- :MFCD00236450
- :316.4610 g/mol
- Conf:25 mg