Orders LinkContactUsLinkComponent
AG-879 ≥99%
AG-879 ≥99%
25 mg
  ENZOBMLEI2580025
AG-879 ≥99%
  ENZOBMLEI2580025
 :  148741-30-4
AG-879 ≥99%
AG-879 ≥99%
25 mg
 :  148741-30-4
SFr. 549.00
1 * 25 mg
NGF receptor inhibitor
AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells.
 :  Tyrphostin AG 879, (E)-2-Cyano-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enethioamide
  •  :148741-30-4
  •  :C₁₈H₂₄N₂OS
  •  :MFCD00236450
  •  :316.4610 g/mol
  • Conf:
    25 mg