Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
AG-879 ≥99%

AG-879 ≥99%

5 mg
Catalog # ENZOBMLEI2580005
Supplier:   Enzo Life Sciences
AG-879 ≥99%
Catalog # ENZOBMLEI2580005
Supplier:  Enzo Life Sciences
CAS Number:  148741-30-4
AG-879 ≥99%
AG-879 ≥99%

5 mg

CAS Number:  148741-30-4
Price
SFr. 135.00
Log in for availability
1 * 5 mg
Product Details & Documents

NGF receptor inhibitor

AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells.

Synonyms:  Tyrphostin AG 879, (E)-2-Cyano-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enethioamide
  • CAS Number:148741-30-4
  • Formula:C₁₈H₂₄N₂OS
  • MDL Number:MFCD00236450
  • Molecular Weight:316.4610 g/mol
Specifications

Specifications

  • Conf:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.