Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Amlodipine ≥98% | Avantor

Amlodipine ≥98%

1 g
Katalognr. ENZOBMLCA2021000
Anbieter:   Enzo Life Sciences
Amlodipine ≥98%
Katalognr. ENZOBMLCA2021000
Anbieter:  Enzo Life Sciences
CAS Nummer:  88150-42-9
Amlodipine ≥98%
Amlodipine ≥98%

1 g

CAS Nummer:  88150-42-9
Preis
340.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Ca²⁺ channel blocker.

Amlodipine is a third generation L-type calcium channel blocker with a unique spectrum of actions, over and above its antihypertensive activity which include stimulation of NO synthase, inhibition of monocyte-endothelial adhesion, inhibition of oxidative stress and sustained duration of action.

Soluble in methanol, DMSO (25 mg/ml), 100% ethanol (25 mg/ml), dimethyl formamide (25 mg/ml) or chloroform.

Synonyme:  3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
  • CAS Nummer:88150-42-9
  • Formel:C₂₀H₂₅ClN₂O₅
  • MDL-Nummer:MFCD00864687
  • Molekulargewicht:408.879 g/mol
  • EINECS:425-820-1
Technische Daten

Ergebnisse des Spezifikationstests

  • Purity:
    ≥98% (Assay)

Technische Daten

  • Conf:
    1 g

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.