Orders LinkContactUsLinkComponent
Genistein
Glass bottle for solids, 25 mg
  ENZOALX350006M025
Genistein
  ENZOALX350006M025
 :  446-72-0
Genistein
Glass bottle for solids, 25 mg
 :  446-72-0
SFr. 99.70
1 * 25 mg
 :  5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
  •  :446-72-0
  •  :C₁₅H₁₀O₅
  •  :MFCD00016952
  •  :270.24 g/mol
  •  :297…298 °C
  •  :207-174-9
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 mg