Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

SB202190 hydrochloride ≥97%

1 mg
Catalog # ENZOALX270268M001
Supplier:   Enzo Life Sciences
SB202190 hydrochloride ≥97%
Catalog # ENZOALX270268M001
Supplier:  Enzo Life Sciences
CAS Number:  350228-36-3
SB202190 hydrochloride ≥97%

1 mg

CAS Number:  350228-36-3
Price
SFr. 167.00
Log in for availability
1 * 1 mg
Product Details & Documents

p38 inhibitor

A water soluble form of the potent p38 (SAPK2a) inhibitor SB202190 (Prod. No. BML-EI294).

Synonyms:  SB-202190 hydrochloride
  • CAS Number:350228-36-3
  • Formula:C₂₀H₁₄FN₃O·HCl
  • Molecular Weight:367.9 g/mol
Specifications

Specifications

  • Conf:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.