Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ac-YVAD-AFC

Ac-YVAD-AFC

5 mg
Codice catalogo ENZOALX260108M005
Nome del fornitore:   Enzo Life Sciences
Ac-YVAD-AFC
Codice catalogo ENZOALX260108M005
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
Ac-YVAD-AFC

5 mg

Numero CAS:  
Price
SFr. 235.00
Log in per disponibilità
1 * 5 mg
Product Details & Documents

Substrate for caspase-1 and -4

Fluorogenic substrate for caspase-1 and -4.

Specifiche

Specifiche

  • Conf:
    5 mg
  • Cat. No.:
    ENZOALX260108M005
  • Protein/peptide name:
    Ac-YVAD-AFC
  • Purezza:
    >99% (HPLC)

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.