Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant FGF-basic 147 | Avantor

Human recombinant FGF-basic 147

50 µG
Katalognr. ENZOALX2018220050
Anbieter:   Enzo Life Sciences
Human recombinant FGF-basic 147
Katalognr. ENZOALX2018220050
Anbieter:  Enzo Life Sciences
CAS Nummer:  
Human recombinant FGF-basic 147

50 µG

CAS Nummer:  
Preis
SFr. 417.00
Einloggen für Verfügbarkeit
1 * 50 µG
Product Details & Documents

Produced in E. coli. Non-glycosylated protein, containing 147 amino acids.

  • Carrier-free
Technische Daten

Technische Daten

  • Conf:
    50 µG
  • Peso molecolare:
    ~16,5kDa
  • Cat. No.:
    ENZOALX2018220050
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF-basic 147
  • Purezza:
    >95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Attività biologica:
    The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Specie:
    Human
  • Formulazione:
    Lyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.

Technische Daten