Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant FGF-basic 147

Human recombinant FGF-basic 147

10 µG
Codice catalogo ENZOALX2018220010
Nome del fornitore:   Enzo Life Sciences
Human recombinant FGF-basic 147
Codice catalogo ENZOALX2018220010
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
Human recombinant FGF-basic 147

10 µG

Numero CAS:  
Price
SFr. 248.00
Log in per disponibilità
1 * 10 µG
Product Details & Documents

Produced in E. coli. Non-glycosylated protein, containing 147 amino acids.

  • Carrier-free
Specifiche

Specifiche

  • Conf:
    10 µG
  • Peso molecolare:
    ~16,5kDa
  • Cat. No.:
    ENZOALX2018220010
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF-basic 147
  • Purezza:
    >95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Attività biologica:
    The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Specie:
    Human
  • Formulazione:
    Lyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.