N-Acetyl-Asp-Glu-OH
50 mg
Codice catalogo ENZOALX151020M050
Nome del fornitore: Enzo Life Sciences
N-Acetyl-Asp-Glu-OH
Codice catalogo ENZOALX151020M050
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Vai a
Product Details & Documents
Glutamate receptor ligand
Endogenous neurotransmitter with high affinity for the brain glutamate receptor. N-acetylated α-linked acidic dipeptidase (NAALADase), a membrane-bound peptidase, hydrolyzes α-NAAG, a major brain peptide, to N-acetylaspartate and glutamate. Discriminates between mGluR2 and mGluR3. mGluR3 activator. NMDA receptor antagonist. Neuroprotective.
Specifiche
Specifiche
- Conf:50 mg
- Cat. No.:ENZOALX151020M050
- Protein/peptide name:N-Acetyl-Asp-Glu-OH
- Purezza:-
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



