Lysates, HeLa (human)
Lysate, HeLa, human, heat shocked, with no recovery time, Pooled, 200 µG
Codice catalogo ENZOADILYCHL102F
Nome del fornitore: Enzo Life Sciences
Lysates, HeLa (human)
Codice catalogo ENZOADILYCHL102F
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Vai a
Product Details & Documents
The HeLa cell line derived from a human donor's epithelioid carcinoma tissue (cervix).
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer.
Heat shock-treated HeLa cells with no recovery time can be a useful tool in studying the regulators of immediate-early gene expression during heat shock stress response.
Specifiche
Specifiche
- Conf:200 µG
- Donatore singolo/associato:Pooled
- Descrizione:Lysate, HeLa, human, heat shocked, with no recovery time
Specifiche
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.