Orders LinkContactUsLinkComponent
Extraction reagents
Extraction reagents
Extraction reagent 2, 5X, 10 mL
Codice catalogo ENZOADI801502
Nome del fornitore:   Enzo Life Sciences
Extraction reagents
Codice catalogo ENZOADI801502
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
Extraction reagents
Extraction reagent 2, 5X, 10 mL
Numero CAS:  
Price
SFr. 208.00
Log in per disponibilità
1 * 10 mL
Product Details & Documents
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.
Specifiche
Specifiche
  • Conf:
    10 mL
  • Descrizione:
    Extraction reagent 2, 5X