Orders LinkContactUsLinkComponent
Viscometers, ROTAVISC SBS
Rotational viscometer package, ROTAVISC SBS hi-vi I Complete, 200 - 80000000 mPas
  IKAA10013820
 :   IKA
Viscometers, ROTAVISC SBS
  IKAA10013820
 :  IKA
Viscometers, ROTAVISC SBS
Viscometers, ROTAVISC SBS
Rotational viscometer package, ROTAVISC SBS hi-vi I Complete, 200 - 80000000 mPas
SFr. 4’980.00
1 * 1 ST
SBS (Steel Ball Bearing, Suspended) rotational viscometers extend a large part of the application spectrum of the IKA ROTAVISC device family.
  • Easy to use
  • Highest measurement accuracy
  • Infinitely variable speed
Instead of jewel bearing, these devices are equipped with a special ball bearing technology.
They are characterised by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
Whether simple or demanding viscosity measurements - with ROTAVISC you get fast and accurate results. Intuitive menu navigation awaits you via the generous 4,3" TFT display. Digital levelling also supports the correct setting of the starting condition. Recurring tasks are facilitated by the simple ramp function.
The accuracy of the ROTAVISC rotary viscometers for Newtonian and non Newtonian liquids is ±1 % of the measuring range. Reproducibility is ±0,2%. With ROTAVISC you have a viscometer in which you can adjust the speed steplessly, but also specify programs or individual speed levels.
 : Complete packages include ROTAVISC SBS (hi-vi I, hi-vi II, or me-vi control), ROTASTAND, SP set-2, PT 100.8, and Guard rail.
HELI complete packages include ROTAVISC SBS (hi-vi I, hi-vi II, or me-vi control), HELISTAND, T-SP-SET, SP set-2, PT 100.8, and Guard rail.
  • Modello:
    ROTAVISC SBS hi-vi I Complete
  • Range di misura:
    200 - 80000000 mPas
  • Descrizione:
    Rotational viscometer package
  • Seq:
    16
  • Cat. No.:
    IKAA10013820
  • Peso:
    7,1 kg
  • L×P×A:
    351×372×629 mm
  • Display:
    TFT
Accessories
  •  
    Descrizione
  • IKAA20116428
    Stand for ROTAVISC
    1 * 1 ST - SFr. 1’200.00
  •  
    Descrizione
  • IKAA25000434
    Olio di silicone standard, CAL-O-100
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25000467
    Standard silicone oil, CAL-O-30000
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25000436
    Olio di silicone standard, CAL-O-1000
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25000468
    Standard silicone oil, CAL-O-60000
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA20128647
    HELISTAND set includes T-spindles for non-flowing samples, EU plug
    1 * 1 ST - SFr. 2’120.00
  •  
    Descrizione
  • IKAA25000352
    Adattatore, VOLS-1, per misurare la viscosità di campioni particolarmente preziosi e costosi
    1 * 1 SET - SFr. 1’680.00
  •  
    Descrizione
  • IKAA25006849
    VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 ml
    1 * 1 ST - SFr. 465.00
  •  
    Descrizione
  • IKAA25000437
    Standard silicone oil, CAL-O-5000
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25000397
    Olio di silicone standard, CAL-O-5
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25006851
    VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 ml
    1 * 1 ST - SFr. 478.00
  •  
    Descrizione
  • IKAA25000466
    Standard silicone oil, CAL-O-12500
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25000435
    Olio di silicone standard, CAL-O-500
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
    Per
  • IKAA20017366
    Labworldsoft® 6 Pro, with access to the entire device library
    All configurations with any number of devices can be controlled at the same time
    1 * 1 ST - SFr. 15’210.00
  •  
    Descrizione
  • IKAA25000390
    Adattatore ELVAS-1 per misurare viscosità molto basse
    1 * 1 SET - SFr. 1’960.00
  •  
    Descrizione
  • IKAA25006852
    VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 ml
    1 * 1 ST - SFr. 478.00
  •  
    Descrizione
    Per
  • IKAA4569000
    Hose, silicone, 2×1,5 m, Ø 8 mm
    1 * 1 ST - SFr. 62.90
  •  
    Descrizione
  • IKAA20114188
    VOLS-1 flex adapter set for low volumes
    1 * 1 ST - SFr. 855.00
  •  
    Descrizione
  • IKAA25000398
    Olio di silicone standard, CAL-O-10
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
    Per
  • IKAA20019397
    Labworldsoft® 6 Starter
    Small configurations with up to 3 devices can be controlled at the same time
    1 * 1 ST - SFr. 6’670.00
  •  
    Descrizione
    Tipo
    Cost. temp.
  • IKAA25004524
    HRC 2 temperature control
    HRC 2 control
    0,05±K
    1 * 1 ST - SFr. 7’770.00
  •  
    Descrizione
  • IKAA25000387
    Adatatore, DINS-1, conforme alla norma DIN 53019
    1 * 1 SET - SFr. 2’960.00
  •  
    Descrizione
  • IKAA25000469
    Standard silicone oil, CAL-O-100000
    1 * 500 mL - SFr. 194.00
  •  
    Descrizione
  • IKAA25006850
    VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 ml
    1 * 1 ST - SFr. 478.00
  •  
    Descrizione
    Per
  • IKAA20101872
    Labworldsoft® 6 Visc
    Viscosity measurements for thermostats and pH meters of all manufacturers
    1 * 1 ST - SFr. 1’490.00
  •  
    Descrizione
  • IKAA25000377
    Adattatore a spirale, SAS-1, per misurare la viscosità di campioni molto appiccicosi e viscosi
    1 * 1 SET - SFr. 1’560.00
  •  
    Descrizione
  • IKAA20118239
    VOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mm
    1 * 1 ST - SFr. 478.00
  •  
    Descrizione
  • IKAA25000407
    Mandrini a palette (set da 4), VAN-1, consentono di misurare la viscosità delle sospensioni
    1 * 1 SET - SFr. 1’490.00
  •  
    Descrizione
  • IKAA25000399
    Olio di silicone standard, CAL-O-50
    1 * 500 mL - SFr. 194.00