Kontermutter PPSU-schwarz für Rührwellenhalter Gleitring RV16 1 * 1 items
Catalog # HWSA3792001/KM-U
Supplier: HWS LABORTECHNIK
Kontermutter PPSU-schwarz für Rührwellenhalter Gleitring RV16 1 * 1 items
Catalog # HWSA3792001/KM-U
Supplier: HWS LABORTECHNIK
Frequently Bought Together
Forceps, sharp
Supplier: BOCHEM
Dichtung, 40x34, PTFE 1 * 1 ST
Supplier: JUCHHEIM Laborgerate GmbH
Magnetic Stirring Bars, PTFE, cylindrical, with pivot ring, BOLA
Supplier: Bohlender
[EN]LARGE CHEMIST SET 9 PCS 1 * 1 pezzi
Supplier: HAMMACHER KARL
FORCEP DUMONT 5SPSF 1 * 1 items
Supplier: Roth Carl
DOPPIO CLASPING IN ACCIAIO INOSSIDABILE 1 * 1 pezzi
Supplier: FINE SCIENCE TOOLS
Fermi per giunti smerigliati conici standard, KECK™, DURAN®
Supplier: DWK Life Sciences
VWR®, Ancorette magnetiche, micro
Supplier: VWR Collection
TREPHINE ACCIAIO INOSSIDABILE 2.7MM DIA2.3MM 1 * 1 pezzi
Supplier: FINE SCIENCE TOOLS
VWR®, Setacci per analisi, 200×50 mm
Supplier: VWR Collection
Test sieves
Supplier: NEXOPART
Vannas spring scissors, 2 mm cutting edge
Supplier: FINE SCIENCE TOOLS
Sterilisation Boxes for Pipettes
Supplier: GLW
Bowls
Supplier: BOCHEM
VWR®, Test Sieves, 400×65 mm
Supplier: VWR Collection