Standard hazard warning signs
ISO Safety sign - Warning, corrosive substance, In PE, 50×43 mm
Codice catalogo BRDY829721
Nome del fornitore: Brady
Vai a
Product Details & Documents
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
- Laminated polyester
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Good weatherability and UV resistance
- Excellent adhesion to low surface energy surfaces
- Thickness: 0,1 mm
Specifiche
- Descrizione:ISO Safety sign - Warning, corrosive substance
- Dimensioni:50×43 mm
- Materiale:In PE
- Seq:03



