MagaDye™ 588 ddTTP
MagaDye™ 588-ddTTP, 5 Nmo
AATB17061
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
- In-house quality control
- Useful for sanger sequencing applications
Caution: For research use only.
- Conf:5 Nmo
- Descrizione:MagaDye™ 588-ddTTP