Orders LinkContactUsLinkComponent
MagaDye™ 613 ddCTP
MagaDye™ 613 ddCTP
MagaDye™ 613-ddCTP, 5 Nmo
Codice catalogo AATB17060
Nome del fornitore:   AAT Bioquest
MagaDye™ 613 ddCTP
Codice catalogo AATB17060
Nome del fornitore:  AAT Bioquest
Numero CAS:  
MagaDye™ 613 ddCTP
MagaDye™ 613 ddCTP
MagaDye™ 613-ddCTP, 5 Nmo
Numero CAS:  
Price
SFr. 1’440.00
Log in per disponibilità
1 * 5 Nmo
Product Details & Documents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Attenzione: For research use only.
Specifiche
Specifiche
  • Conf:
    5 Nmo
  • Descrizione:
    MagaDye™ 613-ddCTP