ContactUsLinkComponent
5 components: 100mg/l each of 4-n-nonylphenol [CAS:104-40-5] 4-Nonylphenol [CAS:84852-15-3] Octylphenol [CAS:27193-28-8] 4-tert-Octylphenol [CAS:140-66-9] 4-n-Octylphenol [CAS:1806-26-4] in Methanol CRM, I 17025 Conservation 12 months. Store 1 * 5 Ampoll
Codice catalogo CMPLRD1036391.5
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
5 components: 100mg/l each of 4-n-nonylphenol [CAS:104-40-5] 4-Nonylphenol [CAS:84852-15-3] Octylphenol [CAS:27193-28-8] 4-tert-Octylphenol [CAS:140-66-9] 4-n-Octylphenol [CAS:1806-26-4] in Methanol CRM, I 17025 Conservation 12 months. Store 1 * 5 Ampoll
Codice catalogo CMPLRD1036391.5
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
Frequently Bought Together