Orders LinkContactUsLinkComponent
Goat serum
Goat serum
Goat serum, heat inactivated, Sterile filtered, Frozen
Codice catalogo S200H-100
Nome del fornitore:   Biowest
Goat serum
Codice catalogo S200H-100
Nome del fornitore:  Biowest
Goat serum
Goat serum
Goat serum, heat inactivated, Sterile filtered, Frozen
Price
SFr. 37.90
Log in per disponibilità
1 * 100 mL
Product Details & Documents
Goat serum is a blood derived product collected from healthy non immunised animals.
  • Blocking agent or negative control in immunoassays
  • For immunohistochemistry and immunocytochemistry, used between 1 to 20% as a blocking agent in various studies
Suitable alternative to FBS in a wide range of cell culture applications (e.g. isolation and identification of viruses produced in a variety of primary cells).
Documents
Specifiche
  • Formato del siero:
    Frozen
  • Sterilità:
    Sterile filtered
  • Descrizione:
    Goat serum, heat inactivated
  • Seq:
    02