Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant FOXA1

Human Recombinant FOXA1

20 µG
Catalog # ORIGTP306045
Supplier:   OriGene
Human Recombinant FOXA1
Catalog # ORIGTP306045
Supplier:  OriGene
CAS Number:  
Human Recombinant FOXA1

20 µG

CAS Number:  
Price
SFr. 1’040.00
Log in for availability
1 * 20 µG
Product Details & Documents

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.

  • Expression host: HEK293T
  • Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol

Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.

Caution: For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.

Specifications

Specifications

  • Conf:
    20 µG
  • Peso molecolare:
    49 kDa
  • Cat. No.:
    ORIGTP306045
  • Condizioni di conservazione:
    Store at −80 °C
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FOXA1
  • Purezza:
    >80% as determined by SDS-PAGE and Coomassie blue staining
  • Protein synonyms:
    HNF3A|forkhead box A1|TCF3A
  • UniProtKB:
    P55317
  • Specie:
    Human
  • Sequenza:
    MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYAN<br />PGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSP<br />MAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQN<br />QQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGS<br />GSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELK<br />TPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAY<br />EQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTS<br /><br />TRTRPLEQKLISEEDLAANDILDYKDDDDKV
  • Concentrazione:
    >0,05 µg/µl as determined by microplate BCA method
  • Sequenza tag:
    C-Myc/DDK

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.