ContactUsLinkComponent
PLEXON OPT/PC-LC-LCF-200/230-HD-1.0L-PD KIT PLEXBRIGHT OPTOGENETIC STIMULATION SYSTEM PATCH CABLE KIT. 200/230µM HIGH DURABILITY FIB ER LC CONNECTOR LC FERRULE WITH POLISHED FIBER ON DISTAL END 1.0M LENGTH . INCLUDES PATCH CABLE 1 * 1 ST
Catalog # MSPP94046-100
PLEXON OPT/PC-LC-LCF-200/230-HD-1.0L-PD KIT PLEXBRIGHT OPTOGENETIC STIMULATION SYSTEM PATCH CABLE KIT. 200/230µM HIGH DURABILITY FIB ER LC CONNECTOR LC FERRULE WITH POLISHED FIBER ON DISTAL END 1.0M LENGTH . INCLUDES PATCH CABLE 1 * 1 ST
Catalog # MSPP94046-100
Frequently Bought Together