VWR® Drawer Racks
Drawer rack, compartment dimensions: 133,0×137,4×75,9 mm, 3×4 compartments, 140×566×256 mm
Codice catalogo 479-2469
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
Specifiche
- Scomparti:3×4 compartments
- Esterno L×P×A:140×566×256 mm
- Descrizione:Drawer rack, compartment dimensions: 133,0×137,4×75,9 mm
- Seq:01



