ContactUsLinkComponent
MOLECULAR INSTRUMENTS BUNDLE 3 HCR BUNDLE HCR CUSTOM PROBE SET ORGANISM: OTHER: HYDRA VULGARIS, PROBE SET CUSTOM: NOGGIN, PROBE SET SIZE: 20, HCR AMPLIFIER: B3, PROBE SET SCALE: 200 PMO
Codice catalogo MSPPOF9ECN5GFXZ-3
Nome del fornitore:  MARKET SOURCE PART PROCESS
MOLECULAR INSTRUMENTS BUNDLE 3 HCR BUNDLE HCR CUSTOM PROBE SET ORGANISM: OTHER: HYDRA VULGARIS, PROBE SET CUSTOM: NOGGIN, PROBE SET SIZE: 20, HCR AMPLIFIER: B3, PROBE SET SCALE: 200 PMO
Codice catalogo MSPPOF9ECN5GFXZ-3
Nome del fornitore:  MARKET SOURCE PART PROCESS
Frequently Bought Together