Krytech 643 Cut Resistant Gloves
Product Details & Documents
KryTech 643 is comfort, suppleness and breathability without any compromise on cut protection and durability can be used with touch devices and touch screen.
- A seamless plaited glove for a maximum comfort, dexterity and flexibility while providing optimal cut protection
- Extended use guaranteed by our exclusive process RESICOMFORT
- Free of harmful substances: OEKO-TEX® standard 100, DMF free
- Interior finish seamless textile support from HDPE fibres
- Exterior finish reinforced grip
- Dermatologically approved
- Gauge: 15
It can be used with touch devices and touch screen. Silicone free to prevent traces, defects on metal sheet and glass prior to painting. Washable one time at 40 °C without any alteration on performances*
* Tests carried out on unworn gloves which differs from the reality in the field.
: ISO certified. This product is not classified hazardous according to the regulation (EC) n°1272/2008 of the European Parliament and of the council. This product does not contain more than 0.1 % of substance of very high concern (SVHC) or any substance included in the annex XVII of the regulation n° 1907/2006 of the European Parliament and of the Council (REACH). UE type certificate or CE type examination certificate: 0075/014/162/10/20/2586. Issued by the notified body nr: 0075 CTC - 4, rue Hermann Frenkel - 69367 Lyon. Cedex 07- FRANCE.
Specifiche
- Dimensione del guanto:7
- Confezione:12 pair/bag
- Colore:nero/grigio
- Lunghezza:230 - 280 mm
- Seq:02
- Modello:Krytech 643
- Cat. No.:MAPA34643017
- Ambidestro/mano specifico:mano specifico



