Labels for BBP33 printers, type B-499
Printer labels, B-499, B33-254-499, Nylon, Acrilico permanente, bianco, BBP33, 19,0×19,0 mm, 1000/roll
BRDY153088
: Brady
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
- Stampante:BBP33
- Tipo:B-499
- Colore:bianco
- Descrizione:Printer labels
- Dimensioni:19,0×19,0 mm
- Materiale:Nylon
- Seq:11
- Modello:B33-254-499
- Confezione:1000/roll
- Adesivo:Acrilico permanente