Orders LinkContactUsLinkComponent
Code Corporation Barcode Reader Accessories
Cable, black, 2,4 m cable length, code readers
Catalog # BRDY176700
Supplier:   Brady
Code Corporation Barcode Reader Accessories
Catalog # BRDY176700
Supplier:  Brady
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories
Cable, black, 2,4 m cable length, code readers
Price
24.10 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Accessories available for barcode readers.
Use the 1,8 m (BRDY176701) or 2,4 m (BRDY176700) coiled RS232 Affinity® cable CRA-C511 with code's cabled barcode readers.
Model BRDY176702 code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3,3 A).
Model BRDY176703 code power supply is an international power supply with USB connection.
Specifications
  • Descrizione:
    Cable, black, 2,4 m cable length, code readers
  • Seq:
    02