Code Corporation Barcode Reader Accessories
Power supply, black, 38,1×76,2×63,5 mm, 6,0 m cable length, CR2700 Quad bay battery charger (BRDY176520)
Codice catalogo BRDY176703
Nome del fornitore: Brady
Vai a
Product Details & Documents
Accessories available for barcode readers.
Use the 1,8 m (BRDY176701) or 2,4 m (BRDY176700) coiled RS232 Affinity® cable CRA-C511 with code's cabled barcode readers.
Model BRDY176702 code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3,3 A).
Model BRDY176703 code power supply is an international power supply with USB connection.
Specifiche
- Descrizione:Power supply, black, 38,1×76,2×63,5 mm, 6,0 m cable length, CR2700 Quad bay battery charger (BRDY176520)
- Seq:04