Cerex® PSAXS SPE Columns
Column Cerex PSAXS, PSAX-S, 300 Å, 10 µm, 1 ml, 50 mg
Catalog # TCAT606-0501R
Supplier: Tecan
Jump to:
Product Details & Documents
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
- Ion-exchange
- Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
Specifications
- Dimensioni particelle:10 µm
- Dimensioni pori:300 Å
- Capacità:1 ml
- Descrizione:Column Cerex PSAXS
- Seq:01
- Peso letto:50 mg
- Fase:PSAX-S