Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
SPE Columns, NBE HPSCX

SPE Columns, NBE HPSCX

Column NBE HPSCX, HPSCX, 30 µm, 1 ml, 2,5 mg
Codice catalogo TCAT427-0021R-NBE
Nome del fornitore:   Tecan
SPE Columns, NBE HPSCX
Codice catalogo TCAT427-0021R-NBE
Nome del fornitore:  Tecan
SPE Columns, NBE HPSCX

Column NBE HPSCX, HPSCX, 30 µm, 1 ml, 2,5 mg

Price
Richiedi prezzo
Log in per disponibilità
1 * 96 ST
Product Details & Documents

Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.

  • Ion-exchange
  • Liquid handler compatible

Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.

Specifiche
  • Dimensioni particelle:
    30 µm
  • Capacità:
    1 ml
  • Descrizione:
    Column NBE HPSCX
  • Seq:
    01
  • Peso letto:
    2,5 mg
  • Fase:
    HPSCX
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.