Orders LinkContactUsLinkComponent
Resolvex® M10 Manual Positive Pressure Processor
Resolvex® M10 Manual Positive Pressure Processor
Resolvex® M10 96 1 ml/NBE columns instrument, 259×305×390 mm
Catalog # TCAT288-0001
Supplier:   Tecan
Resolvex® M10 Manual Positive Pressure Processor
Catalog # TCAT288-0001
Supplier:  Tecan
Resolvex® M10 Manual Positive Pressure Processor
Resolvex® M10 96 1 ml/NBE columns instrument, 259×305×390 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Benchtop manual positive pressure processor.
  • Individual flow restrictors ensure parallel processing even when some wells are vacant
  • Only a gas supply is needed for operation
  • Dual-level gas delivery system offers fine controls of flow and pressure
Perfect for entry into positive pressure processing and workflows, the intuitive design of the Resolvex® M10 makes it easy to use with minimal training required.
Specifications
  • Descrizione:
    Resolvex® M10 96 1 ml/NBE columns instrument, 259×305×390 mm
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0023
    SPE column rack for 6 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0021
    SPE column rack for 1 ml cerex columns, aluminum, W×D×H: 10,8×3,6×3,1"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0022
    SPE column rack for 3 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT253-0006
    Partial plate holder for 1 ml columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT253-0036-SP
    Waste barrier insert (long), plastic, W×D×H: 4,4×3,0×1,6"
    Log in for availability
    1 * 5 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0015
    Collection rack for 12×75 mm tubes for 48 place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT253-0007
    Partial plate holder for NBE columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0016
    Collection rack for 16×100 tubes for 48 mm place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0017
    Collection rack for 12×32 mm autosampler vials for 48 place systems, stainless steel, W×D×H: 10,7×3,5×3,8"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT253-2003
    Mat Luer seal pull tab silicone, H=5,11 mm
    Log in for availability
    1 * 5 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT279-0018
    Collection rack to fit 12×32 mm autosampler vials, stainless steel, W×D×H: 10,7×3,5×3,9"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT700-4009
    Reservoir 96 well 300 ml pyramid, plastic, W×D×H: 5,0×3,4×1,7"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT278-0060
    Mat 96 well pre-slit silic BlankSilicone, H=5,11 mm
    Log in for availability
    1 * 10 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT700-4008
    Plate V-bottom solid 5 ml, plastic, W×D×H: 5,0×3,4×0,6"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219826
    Rack sample 6CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219827
    Rack sample 3CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219825
    Rack collection tube 12×75 mm assay, plastic, W×D×H: 5,0×3,6×3,2"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219821
    Rack vial autosampler assay, plastic, W×D×H: 5,3×3,6×1,7"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219819
    Rack MN giga assay, plastic, W×D×H: 5,0×3,4×4,4"
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • TCAT30219818
    Rack QIAGEN-tip mega assay, plastic, W×D×H: 5,0×3,4×4,3"