BAI1 is a selective and allosteric inhibitor of BAX, an apoptosis regulator. BAI1 directly binds to BAX and allosterically inhibits BAX activation. BAI1 has the potential for the research of diseases mediated by BAX-dependent cell death
Catalog # MDEXHY-103269-10
Supplier: MEDCHEMEXPRESS
Frequently Bought Together